Learn More
Aviva Systems Biology Corp TIMP2 Antibody - middle region (ARP59181_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59181_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TIMP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TIMP2. Alternate names: CSC-21K, Timp-2, Metalloproteinase inhibitor 2, TIMP2, TIMP metallopeptidase inhibitor 2, Tissue inhibitor of metalloproteinases 2, Timp2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-286-609