Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7078RBP1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: Polylconal. Clonality: Monoclonal. Host: Rabbit. Isotype: IgG. Positive Control: Placenta or breast carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: ImmunogenImmunogen for TIMP3 Ab, is 100% homologous in human, cow, Canine, horse,, 94% homologous in mouse, rat. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 30kDa.Chromosome: 22q12.3. Gene Symbol: TIMP3. Gene ID: 7078. Ensemble: ENSG00000100234. Uniprot: NP_000353.
Catalog No. 50-188-5223