Learn More
Aviva Systems Biology Corp TIMP3 Antibody - middle region (ARP59182_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59182_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TIMP3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TIMP3. Alternate names: HSMRK222, K222, K222TA2, SFDApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-286-610