Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TIMP4 ELISA Kit

Catalog No. EHTIMP4
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHTIMP4 96 Tests
EHTIMP4X5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHTIMP4 Supplier Invitrogen™ Supplier No. EHTIMP4
Add to Cart
Add to Cart

ELISA

The Human Tissue Inhibitor of MetalloProteinases 4 (TIMP-4) ELISA quantitates Hu TIMP-4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TIMP-4. Principle of the method The Human TIMP-4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Tissue inhibitor of metalloproteinase 4 (TIMP4) is encoded by the gene TIMP4 located on chromosome 3. It belongs to the family of TIMP genes and is known to inhibit the activity of matrix metalloproteinases. Matrix metalloproteinases (MMP) are a group of proteinases that degrade the components of extracellular matrix. TIMP4 irreversibly binds to MMPs and inactivates them with the help of zinc as a cofactor. The netrin domain of TIMP4 is involved in platelet recruitment and aggregation. Other biological functions include response to cytokine and peptide hormone stimuli and involvement with the ovulation cycle and CNS development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99727
Assay Range 25-7000 pg/mL
Assay Sensitivity 25 pg/mL
Calibration Range 25 pg/mL to 7000 pg/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA- 78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL- 11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF- 7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Sufficient For 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Metalloproteinase inhibitor 4; TIMP 4; TIMP metallopeptidase inhibitor 4; TIMP4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4Metalloproteinase inhibitor 4; TIMP 4; TIMP metallopeptidase inhibitor 4; TIMP4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4;
Gene ID (Entrez) 7079
Gene Symbol TIMP4
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Tissue inhibitor of metalloproteinases 4, TIMP-4
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Development
Sample Volume Plasma 20 μL, Serum 20 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym N/A
Target TIMP4
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.