Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Tin
Spécifications
Spécifications
| Melting Point | 231.9°C |
| CAS | 7440-31-5 |
| Boiling Point | 2270°C |
| Density | 7.310 g/mL at 25°C |
| Quantity | 500 g |
| Molecular Formula | Sn |
| Linear Formula | Sn |
| MDL Number | MFCD00133862 |
| Molecular Weight (g/mol) | 118.71 g/mol |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'