Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin arsenide, 99.999% (metals basis), Thermo Scientific™

Catalog No. AA3636314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3636314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3636314 Supplier Thermo Scientific Chemicals Supplier No. 3636314
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39332-13-3
Molecular Formula C8H10N4O2S
Molecular Weight (g/mol) 226.254
MDL Number MFCD00148646
InChI Key OPKFIKAYNXEJQA-UHFFFAOYSA-N
Synonym 1,2-propanediol, 3-9h-purin-6-ylthio, 3-9h-purin-6-ylsulfanyl propane-1,2-diol, ncistruc1_000545, ncistruc2_000494, 1, 3-purin-6-ylthio, ax 3, 1, 3-1h-purin-6-ylthio, 6-2,3-dihydroxypropyl thiopurine, 3-9h-purin-6-ylthio-1,2-propanediol, 3-7h-purin-6-ylsulfanyl propane-1,2-diol
PubChem CID 3579846
IUPAC Name 3-(7H-purin-6-ylsulfanyl)propane-1,2-diol
SMILES C1=NC2=C(N1)C(=NC=N2)SCC(CO)O

Specifications

Physical Form Pieces
Quantity 25 g
UN Number UN1557
Solubility Information Insoluble in water. Slightly soluble in tin telluride
Formula Weight 193.61
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Tin arsenide
Hazard Category H301+H331
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.