Learn More
Electron Microscopy Sciences SEM Tin On Carbon Medium Resolution Test Specimen, JEOL Stub
SDP
Catalog No. 5029318
Encompass_Preferred
Tin on Carbon is an alternate test specimen for medium resolution, and for the daily basic checking of your instrument performance. This specimen consists of a dispersion of tin spheres, within the size range 10 - 40 nm, on a carbon substrate. Ideal for astigmatism correction, it is also recommended for use in SEM's employed in the semi-conductor industry where the usual gold on carbon specimen cannot be used because of the risk of gold poisoning.
Type: JEOL Stub
Catalog No. 50-293-18 Supplier Electron Microscopy Sciences Supplier No. 7951502
Tin on Carbon is an alternate test specimen for medium resolution, and for the daily basic checking of your instrument performance. This specimen consists of a dispersion of tin spheres, within the size range 10 - 40 nm, on a carbon substrate. Ideal for astigmatism correction, it is also recommended for use in SEM's employed in the semi-conductor industry where the usual gold on carbon specimen cannot be used because of the risk of gold poisoning.
Type: JEOL Stub