Learn More
TA Instruments Tin Calibration Material Not Certified, Standard Tin, Temperature Calibration, DSC Cells, TMA Instruments, Non Certified
Catalog No. 5000580666 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Tin Calibration Material Not Certified is standard tin material used for temperature calibration of DSC cells and TMA instruments where a non-certified reference is sufficient.
Catalog No. 50-005-80666 Supplier TA Instruments Supplier No. 900910.901
Waters TA Instruments Tin Calibration Material Not Certified is standard tin material used for temperature calibration of DSC cells and TMA instruments where a non-certified reference is sufficient.