Learn More
Perkin Elmer US LLC Tin Calibration Standard - 231.88°C
Catalog No. 502113489
Encompass_Preferred
PerkinElmer reference standards are 99.999% pure. DSC instruments requires elements of this purity to ensure accurate calibration. Depend on PerkinElmer for quality calibration reference standards.
Catalog No. 50-211-3489 Supplier Perkin Elmer US LLC Supplier No. 03190034
PerkinElmer reference standards are 99.999% pure. DSC instruments requires elements of this purity to ensure accurate calibration. Depend on PerkinElmer for quality calibration reference standards.