Learn More
Costech Analytical Technologies Inc TIN CAPSULE 5X9 250/PK
Catalog No. nc9914658
Encompass
Tin capsules for solid samples,Size: 5x9 mm,Consumables for the ECS 4010(Elemental Combustion System),NA 1500 Series 2,EA 1108,NA 2500 and EA 1110 Analyzers,Flash EA 1112,NA 1500 Series 1 Analyzer,EA 1106 Analyzer,Europa ANCA N/C Analyzer
Catalog No. NC9914658 Supplier Costech Analytical Technologies Inc Supplier No. 041077
Tin capsules for solid samples,Size: 5x9 mm,Consumables for the ECS 4010(Elemental Combustion System),NA 1500 Series 2,EA 1108,NA 2500 and EA 1110 Analyzers,Flash EA 1112,NA 1500 Series 1 Analyzer,EA 1106 Analyzer,Europa ANCA N/C Analyzer