Learn More
Costech Analytical Technologies Inc TIN CAPSULES 4 X 6 MM 250/PK
Catalog No. NC9601238
Encompass
Tin capsules for solid samples, Size: 4 x 6 mm,Consumables for the ECS 4010(Elemental Combustion System),NA 1500 Series 2,EA 1108,NA 2500 and EA 1110 Analyzers,Flash EA 1112,NA 1500 Series 1 Analyzer,EA 1106 Analyzer,Europa ANCA N/C A
Catalog No. NC9601238 Supplier Costech Analytical Technologies Inc Supplier No. 041070
Tin capsules for solid samples, Size: 4 x 6 mm,Consumables for the ECS 4010(Elemental Combustion System),NA 1500 Series 2,EA 1108,NA 2500 and EA 1110 Analyzers,Flash EA 1112,NA 1500 Series 1 Analyzer,EA 1106 Analyzer,Europa ANCA N/C A