Learn More
Electron Microscopy Sciences Low Magnification SEM Test Specimen Tin On Carbon, Hitachi Stub
SDP
Catalog No. 501931220
Encompass_Preferred
This low magnification test has a size range of 1 - 10μm for table-top SEM. Has a magnification range of 250-5,000x. Can be used on FESEM and FIB as well.
Type: Hitachi Stub
Catalog No. 50-193-1220 Supplier Electron Microscopy Sciences Supplier No. 7951804
This low magnification test has a size range of 1 - 10μm for table-top SEM. Has a magnification range of 250-5,000x. Can be used on FESEM and FIB as well.
Type: Hitachi Stub