Learn More
Crescent Chemical Co Inc TIN ICP STD 100ML
SDP
Catalog No. 5072138
Encompass_Preferred
Tin, HCL Single Element Plasma Emission Spectroscopy Standards 1,000 UgmL NIST Traceable CAS Size - 100mL Catalog Number - 8624
Catalog No. 50-721-38 Supplier Crescent Chemical Co Inc Supplier No. IS8624 (100ML)
Add to Cart
Add to Cart
Tin, HCL Single Element Plasma Emission Spectroscopy Standards 1,000 UgmL NIST Traceable CAS Size - 100mL Catalog Number - 8624