Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tin(II) 2-Ethylhexanoate

Catalog No. S3252250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S3252250G 1 kg
S32521KG 100 g
S3252100G 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. S3252250G Supplier Sigma Aldrich Supplier No. S3252250G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tin(II) 2-ethylhexanoate is commonly used as a catalyst for the ring-opening polymerization of cyclic diesters such as L-lactide and glycolide to form high-molecular-weight polymers.

Specifications

CAS 301-10-0
Density 1.251 g/mL at 25°C
Molecular Formula C16H30O4Sn
Linear Formula [CH3(CH2)3CH(C2H5)CO2]2Sn
Refractive Index n20/D 1.493
MDL Number MFCD00002676
Quantity 1 kg
Synonym 2-Ethylhexanoic acid tin(II) salt; Stannous 2-ethylhexanoate; Stannous octoate
Molecular Weight (g/mol) 405.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.