Learn More
Strem, An Ascensus Company CAS# 638-39-1. 5g. Tin(II) acetate, 99%. MFCD00054408
SDP
Supplier:  Strem, An Ascensus Company 5019755G
Encompass
CAS# 638-39-1. 5g. Tin(II) acetate, 99%. MFCD00054408. Molecular Weight: 236.78. Molecular Formula: Sn(OOCCH3)2. Color/form: off-white pwdr. Strem# 50-1975. http://www.strem.com/catalog/v/50-1975/
Catalog No. 50-901-12609