Learn More
Strem, An Ascensus Company CAS# 10031-24-0. 10g. Tin(II) bromide, 99%. MFCD00011239
SDP
Supplier:  Strem, An Ascensus Company 93502810G
Encompass
CAS# 10031-24-0. 10g. Tin(II) bromide, 99%. MFCD00011239. Molecular Weight: 278.51. Molecular Formula: SnBr2. Color/form: gray pwdr. Strem# 93-5028. http://www.strem.com/catalog/v/93-5028/
Catalog No. 50-901-15580