Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(II) bromide, 99.2%

Catalog No. aa4343014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4343014 25 g
AA4343006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4343014 Supplier Thermo Scientific Chemicals Supplier No. 04343014
Add to Cart

CAS: 10031-24-0 | SnBr2

Stannous bromide acts a lewis acid. It is used to prepare alkyl tin compounds and in the spectrophotometric determination of iridium. It is involved in regioselective mono-etherfications of vicinal diols with diazo compounds. It acts as a catalyst to produce resinous lactone high polymers. It serves as a potent reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-24-0
Molecular Formula SnBr2
MDL Number MFCD00011239
Synonym tin ii bromide, tin dibromide, stannous bromide, tin bromide snbr2, snbr2, acmc-1bp22

Specifications

Boiling Point 623°C
Melting Point 215°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.4%
Solubility Information Soluble in alcohol,ether,acetone,pyridine and dimethyl sulfoxide.
Formula Weight 278.51
Density 5.12
Chemical Name or Material Tin(II) bromide
Hazard Category H302-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 233-087-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.