Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(Ii) Chloride, Solstice

Catalog No. 6001560 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-015-60 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-015-60 Supplier Solstice Supplier No. 2082562KG

missing translation for 'validMainframeMsg'

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
PubChem CID 24479
ChEBI CHEBI:78067
IUPAC Name dichlorotin
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Quantity 2 kg
Linear Formula SnCl2
UN Number 3260
Formula Weight 189.62g/mol
Percent Purity 98%
Grade Reagent
Chemical Name or Material Tin(Ii) Chloride
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314-H317-H332-H335-H341-H361-H373-H410
EINECSNumber 231-868-0
RTECSNumber XP8700000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.