Learn More
Tin(II) chloride, ultra dry, 99.9985% (metals basis)
Description
Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It acts as an antisludging agent and a stabilizer. It can be used to reduce certain metal salts into metals, tin plating and in analytical chemistry to test mercury ions. It is also used as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. It is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 652°C |
| Melting Point | 247°C |
| Physical Form | Powder |
| Quantity | 100 g |
| Sensitivity | Air sensitive; Hygroscopic |
| Assay Percent Range | 99.9%, metal basis |
| Density | 3.95 |
| Solubility Information | Soluble in water,alkalies,alcohol,methyl ethyl ketone,methyl acetate and acetone. |
| IUPAC Name | dichlorotin |
| Formula Weight | 189.62 |
| Show More |
Safety and Handling
| Hazard Category | H290-H302+H332-H314-H317-H335-H373 |
| Hazard Statement | Avoid contact with skin. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). |
| Precautionary Statement | P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c |
| DOTInformation | DOT Class: 8; Packaging Group: III |
| EINECSNumber | 231-868-0 |
| RTECSNumber | XP8700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.