Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(II) ethoxide

Catalog No. p-7049746
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2089603 1 g
AA2089606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2089603 Supplier Thermo Scientific Chemicals Supplier No. 02089603
Only null left
Add to Cart

CAS: 14791-99-2 | C4H10O2Sn | 208.832 g/mol

Tin(II) ethoxide is used in catalyst synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14791-99-2
Molecular Formula C4H10O2Sn
Molecular Weight (g/mol) 208.832
MDL Number MFCD00015653
InChI Key XCKWFNSALCEAPW-UHFFFAOYSA-N
Synonym tin ii ethoxide, diethoxytin ii, acmc-20aji5, ethanol, tin 2+ salt 9ci
PubChem CID 71317418
IUPAC Name ethanolate;tin(2+)
SMILES CC[O-].CC[O-].[Sn+2]

Specifications

Melting Point ∼200°C (decomposition)
Linear Formula Sn(OC2H5)2
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water
Formula Weight 208.81
Physical Form Solid/Semi-solid
Chemical Name or Material Tin(II) ethoxide
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.