Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tin(II) Fluoride

Catalog No. 334626100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-462-6100G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 33-462-6100G Supplier Sigma Aldrich Supplier No. 334626100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tin(II) fluoride (SnF2) can be used:As an inhibitor of Sn4+ in fabrication of formamidinium tin iodide (FASnI3) perovskite solar cells (PSCs) via SnF2-pyrazine complex formation.As a catalyst in glycerol ketalization, Knoevenagel condensation and aldol-type reactions.In the synthesis of +a, +a-epoxy ketone from +a, +a-dibromoacetophenone and aliphatic and aromatic aldehyde via oxidative addition of carbon-halogen bonds.In the stereocontrolled [3 + 3] , [3 + 4], [3 + 5] annulations reactions.

Specifications

CAS 3674-54-2
Melting Point 120°C to 123°C
Molecular Formula C17H36N2S
Linear Formula (CH3CH2CH2CH2)4N(SCN)
MDL Number MFCD00012140
Quantity 10 g
Synonym Difluorostannylene; Stannous fluoride; Tin bifluoride; Tin difluoride
Molecular Weight (g/mol) 300.55 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.