Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(II) fluoride, 97.5%

Catalog No. p-7048814
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1154422 100 g
AA1154436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1154422 Supplier Thermo Scientific Chemicals Supplier No. 01154422
Only null left
Add to Cart

CAS: 7783-47-3

Tin(II) fluoride is used in the prevention of dental caries. It is an active ingredient in toothpastes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-47-3
MDL Number MFCD00042540

Specifications

Boiling Point 850°C
Melting Point 219°C
Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula SnF2
UN Number UN3260
Merck Index 14,8784
Solubility Information Soluble in potassium hydroxide and potassium fluoride. Insoluble in water,ethanol,ether and chloroform.
Formula Weight 156.69
Percent Purity 97.5%
Odor Odorless
Sensitivity Air and moisture sensitive
Density 4.57 g/mL
Chemical Name or Material Tin(II) fluoride
Show More Show Less
Hazard Category H290-H301-H315-H318
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P234-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-999-3
RTECSNumber XQ3450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.