Learn More
Strem, An Ascensus Company CAS# 10294-70-9. 5g. Tin(II) iodide, 99%. MFCD00049545
SDP
Catalog No. 5090115622
Encompass
CAS# 10294-70-9. 5g. Tin(II) iodide, 99%. MFCD00049545. Molecular Weight: 372.50. Molecular Formula: SnI2. Color/form: -6 mesh orange to red pwdr. Strem# 93-5077. http://www.strem.com/catalog/v/93-5077/
Catalog No. 50-901-15622 Supplier Strem, An Ascensus Company Supplier No. 9350775G
CAS# 10294-70-9. 5g. Tin(II) iodide, 99%. MFCD00049545. Molecular Weight: 372.50. Molecular Formula: SnI2. Color/form: -6 mesh orange to red pwdr. Strem# 93-5077. http://www.strem.com/catalog/v/93-5077/