Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tin(II) Methanesulfonate Solution

Catalog No. 462675250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-267-5250ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46-267-5250ML Supplier Sigma Aldrich Supplier No. 462675250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tin(II) Methanesulfonate Solution

Specifications

CAS 90315-82-5
Density 1.075 g/mL at 20°C
Boiling Point 212°C
Molecular Formula C12H16O3
Refractive Index n20/D 1.444
Linear Formula C6H5CH2CH2CH(OH)CO2C2H5
MDL Number MFCD00077794
Quantity 5 g
Synonym Stannous methanesulfonate; Stannous methylsulfonate
Molecular Weight (g/mol) 208.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.