Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tin(II) Sulfate

Catalog No. 244635500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-463-5500G 500 g
24-463-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 24-463-5500G Supplier Sigma Aldrich Supplier No. 244635500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

General applications:Tin(II) sulfate can be used in the electrosynthesis of Cu2ZnSnS4 (CZTS) thin films for solar cell application.It can be used to synthesize ternary Cu-Sn-S chalcogenides such as Cu2SnS3, Cu5Sn2S7 and Cu3SnS4 by successive ionic layer adsorption and reaction (SILAR) method.SnSO4 can also be used in the preparation of tin (IV) oxide (SnO2) nanoparticles which are widely used in battery electrodes, fabrication of supercapacitors and opto-electronic devices.

Specifications

CAS 7488-55-3
Quantity 500 g
Molecular Formula O4SSn
Linear Formula SnSO4
MDL Number MFCD00011246
Synonym Stannous sulfate
Molecular Weight (g/mol) 214.77 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.