Learn More
Strem, An Ascensus Company CAS# 7488-55-3. 100g. Tin(II) sulfate, 95+%. MFCD00011246
SDP
Supplier:  Strem, An Ascensus Company 935044100G
Encompass
CAS# 7488-55-3. 100g. Tin(II) sulfate, 95+%. MFCD00011246. Molecular Weight: 214.75. Molecular Formula: SnSO4. Color/form: white pwdr. Strem# 93-5044. http://www.strem.com/catalog/v/93-5044/
Catalog No. 50-901-15594