Learn More
Strem, An Ascensus Company CAS# 7789-67-5. 25g. Tin(IV) bromide, 99%. MFCD00011240
SDP
Supplier:  Strem, An Ascensus Company 93502125G
Encompass
CAS# 7789-67-5. 25g. Tin(IV) bromide, 99%. MFCD00011240. Molecular Weight: 438.33. Molecular Formula: SnBr4. Color/form: colorless to yellow xtl. Strem# 93-5021. http://www.strem.com/catalog/v/93-5021/
Catalog No. 50-901-15568