Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(IV) Chloride Pentahydrate, 98%, Solstice

Catalog No. 60045632 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-632 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-632 Supplier Solstice Supplier No. 2446781KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

>98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-06-9
Molecular Formula Cl4H10O5Sn
Molecular Weight (g/mol) 350.59
MDL Number MFCD00149864
InChI Key KHMOASUYFVRATF-UHFFFAOYSA-J
Synonym tin iv chloride pentahydrate, tetrachlorostannane pentahydrate, tin chloride pentahydrate, stannic chloride pentahydrate, stannane, tetrachloro-, pentahydrate, ccris 6330, tin iv chloride, pentahydrate 1:4:5, cl4sn.5h2o, stannic chloride 5h2o
PubChem CID 61442
IUPAC Name tin(4+) pentahydrate tetrachloride
SMILES O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]

Specifications

Color White to Yellow
Physical Form Crystalline
Quantity 1 kg
Packaging Poly Bottle
Appearance Powder or Chunks
Linear Formula SnCl4 · 5H2O
UN Number UN2440
Formula Weight 350.6g/mol
Percent Purity 98%
Grade Technical
Chemical Name or Material Tin(IV) Chloride Pentahydrate
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314
DOTInformation 8/PGIII
RTECSNumber XP8870000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.