Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tin(IV) Chloride Solution

Catalog No. 503081490 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1490 4 x 25 mL
24-995-5800ML 800 mL
24-995-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 50-308-1490 Supplier Sigma Aldrich Supplier No. 2499554X25ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tin(IV) chloride shows Lewis acid character and is generally used for the synthesis of SnO2 nanoparticles by sol-gel method.Tin(IV) chloride solution can be used for chloromethylation reaction of polysulfones to develop polymer electrolyte membranes.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 7646-78-8
Density 1.419 g/mL at 25°C
Molecular Formula Cl4Sn
Linear Formula SnCl4
Quantity 4 x 25 mL
MDL Number MFCD00011242
Synonym Stannic chloride; Tin tetrachloride
Molecular Weight (g/mol) 260.52 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.