Learn More
Medchemexpress LLC HY-13707 1mg Medchemexpress, Tin(IV) mesoporphyrin IX (dichloride) CAS:106344-20-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY137071MG
Encompass_Preferred
Medchemexpress, HY-13707 1mg Tin(IV) mesoporphyrin IX (dichloride) CAS:106344-20-1 Tin(IV) mesoporphyrin IX dichloride (Stannsoporfin) is a heme oxygenase (HO) inhibitor being developed for the prevention of hyperbilirubinemia in infants at risk of developing jaundice, extracted from patent WO2011103196A1 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2678