Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(IV) oxide, 15% in H{2}O colloidal dispersion

Catalog No. AA4459236
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4459236 500 g
AA44592A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4459236 Supplier Thermo Scientific Chemicals Supplier No. 04459236

missing translation for 'validMainframeMsg'

Particle charge ± negative | CAS: 18282-10-5 | O2Sn | 150.71 g/mol

It is used to detect carbon monoxide. Used in ceramic glazes and colors, putty, perfume preparations, textiles, polishing powder for steel, and the manufacture of special glasses. Stannic oxide is also used in making white patinas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name stannanedione
SMILES O=[Sn]=O

Specifications

Physical Form Liquid
Quantity 500 g
Assay Percent Range 15% in H2O colloidal dispersion
Solubility Information Fully miscible in water.
Formula Weight 150.69
Density 1.153 g/mL
Chemical Name or Material Tin(IV) oxide, pH 10.5-12.0, Viscosity 5-10cps, Particle charge ±negative. 1.15(Sp Grav)
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.