Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tin(IV) oxide nanoparticle ink

Catalog No. 90107950ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-107-950ML 50 mL
90-107-910ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-107-950ML Supplier Sigma Aldrich Supplier No. 90107950ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This SnO2 nanoparticle ink is for charge selective layers in printed electronics.This SnO2 nanoparticle ink is for spin-coating, doctor blading, slot-die or inkjet printing.Product of Avantama Ltd.

Specifications

Quantity 50 mL
Linear Formula SnO2
Synonym Avantama N-31; Ink for charge selective layers in printed electronics; SnO2 ink; Tin oxide nanoparticle dispersion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.