Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Tin, Metal, Powder
SCI_ED

Catalog No. S25608 Shop All Fisherbrand Products
Change view
Click to view available options
100g; Reagent Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S25608 100g; Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25608

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Gray
CAS 7440-31-5
Density 7.31g/mL
Quantity 100 g
Packaging Reagent Grade - Powder
Molecular Formula Sn
Linear Formula Sn
MDL Number MFCD00133862
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES [Sn]
IUPAC Name tin
Molecular Weight (g/mol) 118.71
PubChem CID 5352426
ChEBI CHEBI:32990
Formula Weight 118.7
Grade Reagent
Chemical Name or Material Tin
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.