Learn More
Apexbio Technology LLC Tin protoporphyrin IX dichloride(Synonyms: Tin Protoporphyrin Dichloride, SnPP, SnPP IX, Tin(IV) Protoporphyrin Dichloride), 25mg, CAS: 14325-05-4.
SDP
Catalog No. 5000060674
Encompass_Preferred
Tin protoporphyrin IX dichloride (CAS 14325-05-4) is a synthetic metalloporphyrin analog that selectively inhibits heme oxygenase-2 (HO-2) an enzyme abundant in neuronal and endothelial tissues responsible for catalyzing heme breakdown into biliverdin and carbon monoxide (CO) It shows selectivity for HO over endothelial nitric oxide synthase and soluble guanylyl cyclase with IC50 values of 35 and 30 nM respectively In rat neonates administration of this compound suppresses the physiological increase in HO activity post-birth and prevents neonatal hyperbilirubinemia demonstrating utility in biomedical research on heme metabolism pathways
Catalog No. 50-000-60674 Supplier Apexbio Technology LLC Supplier No. B643225
Tin protoporphyrin IX dichloride (CAS 14325-05-4) is a synthetic metalloporphyrin analog that selectively inhibits heme oxygenase-2 (HO-2) an enzyme abundant in neuronal and endothelial tissues responsible for catalyzing heme breakdown into biliverdin and carbon monoxide (CO) It shows selectivity for HO over endothelial nitric oxide synthase and soluble guanylyl cyclase with IC50 values of 35 and 30 nM respectively In rat neonates administration of this compound suppresses the physiological increase in HO activity post-birth and prevents neonatal hyperbilirubinemia demonstrating utility in biomedical research on heme metabolism pathways