Learn More
Selleck Chemical LLC Tin protoporphyrin IX
SDP
Catalog No. 5000001629
Encompass_Preferred
Tin Protoporphyrin IX dichloride is a potent inhibitor of heme oxygenase-1 (HO-1) reduces ozone-induced pulmonary inflammation and BAL cell necrosis highlighting its protective role It also sensitizes pancreatic ductal adenocarcinoma (PDAC) tumors to chemotherapy in a mouse model suggesting its potential for PDAC treatment
Catalog No. 50-000-01629 Supplier Selleck Chemical LLC Supplier No. S78795MG
Tin Protoporphyrin IX dichloride is a potent inhibitor of heme oxygenase-1 (HO-1) reduces ozone-induced pulmonary inflammation and BAL cell necrosis highlighting its protective role It also sensitizes pancreatic ductal adenocarcinoma (PDAC) tumors to chemotherapy in a mouse model suggesting its potential for PDAC treatment