Learn More
TARGETMOL CHEMICALS INC Tin-protoporphyrin IX dichlori
SDP
Catalog No. 502876398
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Tin-protoporphyrin IX dichloride (SnPPIX dichloride) is an HO-1) inhibitor with antiviral activity that inhibits dengue and yellow fever viruses. Purity 98%
Catalog No. 50-287-6398 Supplier TARGETMOL CHEMICALS INC Supplier No. T131615MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Tin-protoporphyrin IX dichloride (SnPPIX dichloride) is an HO-1) inhibitor with antiviral activity that inhibits dengue and yellow fever viruses. Purity 98%