Learn More
Strem, An Ascensus Company CAS# 7440-31-5. 250g. Tin powder (99.8%). MFCD00133862
SDP
Catalog No. 5090115639
Encompass
CAS# 7440-31-5. 250g. Tin powder (99.8%). MFCD00133862. Molecular Weight: 118.70. Molecular Formula: Sn. Color/form: -325 mesh Strem# 93-5086. http://www.strem.com/catalog/v/93-5086/
Catalog No. 50-901-15639 Supplier Strem, An Ascensus Company Supplier No. 935086250G
CAS# 7440-31-5. 250g. Tin powder (99.8%). MFCD00133862. Molecular Weight: 118.70. Molecular Formula: Sn. Color/form: -325 mesh Strem# 93-5086. http://www.strem.com/catalog/v/93-5086/