Learn More
Perkin Elmer US LLC Tin (Sn) Pure Standard, 10,000 µg/mL, 20% HCl, 500 mL
Catalog No. 502111783
Encompass_Preferred
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
Catalog No. 50-211-1783 Supplier Perkin Elmer US LLC Supplier No. N9304346
Each Single Element Calibration Standard for AA and ICP-OES is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.