Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampco Tin Snips
SDP

Catalog No. 19174258 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
14 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-174-258 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-174-258 Supplier Ampco Supplier No. S1144A
Add to Cart
Add to Cart

Precision non-sparking, non-magnetic cutting tools designed for safe use in hazardous, explosive, or MRI environements.

  • Independently tested and found to pose no risk in MRI environments using a static magnetic field of 3-Tesla and a maximum spatial gradient magnetic field of 720-Gauss/cm or less.
  • Conforms to ASTM F2503.
  • Meets the requirements established by ASTM A342.
  • Metric blade length: 115 mm
  • Metric overall length: 355 mm
  • English blade length: 4 and 1/2 in
  • English overall length: 14 in.

Specifications

Product Type Tin Snips
Length (English) 14 in.
Length (English) Blade 4 -1/2 in.
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.