Learn More
SPEX CERTIPREP LLC 3P Tin at 1,000 ug/mL in 2% HNO3 + 0.5% HF, 100 mL
Catalog No. NC0911097
Encompass_Preferred
(HPS by Zeptometrix) Tin (Sn) ICP Single-Component Standard at 1,000 ug/mL in 2% HNO3 + 0.5% HF.
Catalog No. NC0911097 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000613100
(HPS by Zeptometrix) Tin (Sn) ICP Single-Component Standard at 1,000 ug/mL in 2% HNO3 + 0.5% HF.