Learn More
Aviva Systems Biology Corp TINAGL1 Antibody - middle region (ARP57621_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57621_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TINAGL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TINAGL1. Alternate names: ARG1, LCN7, LIECG3, TINAGRPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-63027