Learn More
MSC Tingley Bib Overall, Polyurethane/Nylon, Blue, Large, 1 ea
Catalog No. 5000685452
Encompass
These Iron Eagle Bib Overalls are made from coated polyurethane on nylon, offering a lightweight, comfortable, and flexible design. They are engineered to resist fats, oils, pine tars, gasoline, grease, and various other chemicals.
  • Constructed from coated polyurethane on nylon
  • Provides 100% waterproof protection with Therm-O-Rad heat-sealed seams
  • Resistant to fats, oils, and numerous chemicals
  • Provides good resistance to most hydrocarbon oils, organic acids, salts, and alkalies
  • Features a 2 mil exterior coating for snag and abrasion resistance
  • Mildew resistant coating for easy decontamination and liquid shedding
  • Equipped with heavy-duty suspenders and premium snap-lock buckles
  • Breathable, comfortable, and flexible design
Catalog No. 50-006-85452 Supplier MSC Supplier No. 39754049
These Iron Eagle Bib Overalls are made from coated polyurethane on nylon, offering a lightweight, comfortable, and flexible design. They are engineered to resist fats, oils, pine tars, gasoline, grease, and various other chemicals.
  • Constructed from coated polyurethane on nylon
  • Provides 100% waterproof protection with Therm-O-Rad heat-sealed seams
  • Resistant to fats, oils, and numerous chemicals
  • Provides good resistance to most hydrocarbon oils, organic acids, salts, and alkalies
  • Features a 2 mil exterior coating for snag and abrasion resistance
  • Mildew resistant coating for easy decontamination and liquid shedding
  • Equipped with heavy-duty suspenders and premium snap-lock buckles
  • Breathable, comfortable, and flexible design