Learn More
MSC Tingley Bib Overalls, Polyurethane Coated Nylon, Waterproof, 2X-Large, 1 ea
Catalog No. 5000793972
Encompass
These Iron Eagle Overalls are constructed from polyurethane coated nylon, offering durable protection against various elements. Designed for comfort and flexibility, they are suitable for demanding industrial and agricultural environments.
  • Therm-O-Rad sealed seams for 100% waterproof protection
  • Premium snap lock suspender buckles and heavy-duty snaps
  • Mildew resistant with a 2 mil exterior coating for snag and abrasion resistance
  • Sleek coating allows for easy decontamination and shedding of liquids
  • Heat-sealed seams are stronger than the material itself and withstand extensive laundering
  • Resists fats, oils, pine tars, gasoline, grease, and many other chemicals
Catalog No. 50-007-93972 Supplier MSC Supplier No. 63567226
These Iron Eagle Overalls are constructed from polyurethane coated nylon, offering durable protection against various elements. Designed for comfort and flexibility, they are suitable for demanding industrial and agricultural environments.
  • Therm-O-Rad sealed seams for 100% waterproof protection
  • Premium snap lock suspender buckles and heavy-duty snaps
  • Mildew resistant with a 2 mil exterior coating for snag and abrasion resistance
  • Sleek coating allows for easy decontamination and shedding of liquids
  • Heat-sealed seams are stronger than the material itself and withstand extensive laundering
  • Resists fats, oils, pine tars, gasoline, grease, and many other chemicals