Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Detachable Hoods

Catalog No. 19820228 Shop All Tingley Rubber Co. Products
Change view
Click to view available options
Size: large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-820-228 Size: large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-820-228 Supplier Tingley™ Supplier No. H31107.LG
This item is not returnable. View return policy

Specifications

Size Large
Color Yellow
Seam Type Sealed
Thickness 26 mil
For Use With (Equipment) Resistant to mildew, acids, alcohols, salts and alkalis, Waterproof protection
Quantity 1 Ea.
Includes Heavy coating and high-loft lining
Resistance Resistant to mildew, acids, alcohols, salts and alkalis, Waterproof protection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.