Learn More
MSC Tingley Eagle Jacket, 200 Denier Nylon Waterproof Large, 1 ea
Catalog No. 5000688063
Encompass
The Eagle jacket is a breathable polyurethane on nylon jacket designed for all-day comfort and wear. It is the most lightweight and comfortable industrial rain jacket, constructed of 200 denier nylon with an interior polyurethane coating.
  • Raglan sleeve construction for comfortable fit
  • Stitched and taped seams for 100% waterproof protection
  • Storm fly front seals out wind, rain and contaminants
  • Hood snaps compatible with a detachable hood (sold separately)
  • Non-conductive, non-corrosive snaps
  • Mildew resistant
  • Breathable (test method ASTM E96)
  • Generously cut to fit over heavy work clothes
  • Interior polyurethane coating and exterior DWR coating for 100% waterproof protection
Catalog No. 50-006-88063 Supplier MSC Supplier No. 86205895
The Eagle jacket is a breathable polyurethane on nylon jacket designed for all-day comfort and wear. It is the most lightweight and comfortable industrial rain jacket, constructed of 200 denier nylon with an interior polyurethane coating.
  • Raglan sleeve construction for comfortable fit
  • Stitched and taped seams for 100% waterproof protection
  • Storm fly front seals out wind, rain and contaminants
  • Hood snaps compatible with a detachable hood (sold separately)
  • Non-conductive, non-corrosive snaps
  • Mildew resistant
  • Breathable (test method ASTM E96)
  • Generously cut to fit over heavy work clothes
  • Interior polyurethane coating and exterior DWR coating for 100% waterproof protection