Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Electra™ PVC/Nomex™ Overalls

Catalog No. 19820221 Shop All Tingley Rubber Co. Products
Change view
Click to view available options
Fluorescent orange-red; Size: X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-820-221 Fluorescent orange-red; Size: X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-820-221 Supplier Tingley™ Supplier No. O42129.XL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Provide warmth, waterproof protection and excellent visibility

Fluorescent background colors allow for excellent daytime visibility; 2 in. silver reflective trim reflects light for 360° nighttime conspicuity.

  • Light weight with flame-resistant PVC coating on nonwoven Nomex, 9.5 oz., 15mil thick
  • Full-cut shoulder with 90° sleeve to body angle for maximum freedom of movement
  • Therm-O-Rad™ sealed seams for waterproof protection
  • Excellent thermal performance
  • Corduroy collar with hood in collar construction
  • Oversized three-piece hood with adjustable velcro take-up strap
  • Zipper/storm fly front seals out wind, rain, and contaminants
  • ARC Rating, Lime: 7 cal/cm2, HAF: 69%, Break open: 38.6 cal/cm2
  • ARC Rating, Orange: 11 cal/cm2, HAF: 79.2%, Break open: 36.4 cal/cm

  • Non-conductive hardware
  • Full-vented cape back for improved air circulation
  • D-ring access for fall protection harness
  • Heavy duty quick release suspender buckles
  • Easy access cargo pockets with closure
  • Velcro take-up straps for adjustable fit
  • Mildew and chemical resistant

Line workers, troubleshooting, substation, switching, pipeline services, and construction.

Compliance

Conform to ASTM™ F1891 for Arc Thermal Performance. Meets ANSI™/ISEA 107-2010 standards. Class 3 compliant.

Specifications

Closure Type Snap Fly with Hook and Loop Strap
Color Fluorescent Orange, Red
Seam Type Therm-O-Rad Sealed
Size X-Large
Arc Flash Rating 11 Calories/cm2
For Use With (Application) Line workers, trouble shooting, substation, switching, pipeline services and construction
Thickness 15 mil
Certifications/Compliance ANSI ISEA 107 Class E, ASTM F1891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.