Learn More
MSC Tingley Iron Eagle Jacket, 2XL, Green, 1 ea
Catalog No. 5000787061
Encompass
The Iron Eagle jacket features an exterior coated polyurethane on nylon, offering a lightweight yet durable design. It is comfortable, flexible, and provides resistance against fats, oils, pine tars, gasoline, grease, and various other chemicals.
  • Polyurethane on nylon material
  • Batwing sleeve for maximum freedom of movement
  • Therm-O-Rad sealed seams provide 100% waterproof protection
  • Storm fly front seals out wind, rain, and contaminants
  • Hood snaps compatible with Iron Eagle hood (sold separately)
  • Heavy-duty snaps guaranteed not to fail
  • Mildew resistant
  • Sleek coating is easy to decontaminate and effectively sheds liquids
  • Two mil exterior coating offers snag and abrasion resistance
  • Good resistance to most hydrocarbon oils, organic acids, salts, alkalies, and some organic solvents
Catalog No. 50-007-87061 Supplier MSC Supplier No. 81648644
The Iron Eagle jacket features an exterior coated polyurethane on nylon, offering a lightweight yet durable design. It is comfortable, flexible, and provides resistance against fats, oils, pine tars, gasoline, grease, and various other chemicals.
  • Polyurethane on nylon material
  • Batwing sleeve for maximum freedom of movement
  • Therm-O-Rad sealed seams provide 100% waterproof protection
  • Storm fly front seals out wind, rain, and contaminants
  • Hood snaps compatible with Iron Eagle hood (sold separately)
  • Heavy-duty snaps guaranteed not to fail
  • Mildew resistant
  • Sleek coating is easy to decontaminate and effectively sheds liquids
  • Two mil exterior coating offers snag and abrasion resistance
  • Good resistance to most hydrocarbon oils, organic acids, salts, alkalies, and some organic solvents