Learn More
MSC Tingley Iron Eagle Overalls, Polyurethane/Nylon, Waterproof, Small, Green, 1 ea
Catalog No. 5000688425
Encompass
These overalls are crafted from coated polyurethane on nylon, offering a lightweight yet durable design. They provide exceptional comfort and flexibility, and are engineered to resist fats, oils, pine tars, gasoline, grease, and a variety of other chemicals, making them suitable for demanding environments.
  • Therm-O-Rad sealed seams ensure waterproof protection
  • Premium snap lock suspender buckles for secure fit
  • Heavy-duty suspenders with crossover design
  • Heavy-duty snaps resist failure
  • Mildew resistant 2 mil exterior coating
  • Exterior coating provides snag and abrasion resistance
  • Sleek coating is easy to decontaminate and sheds liquids effectively
  • Heat-sealed seams are stronger than the material itself, allowing for extensive laundering
Catalog No. 50-006-88425 Supplier MSC Supplier No. 96549290
These overalls are crafted from coated polyurethane on nylon, offering a lightweight yet durable design. They provide exceptional comfort and flexibility, and are engineered to resist fats, oils, pine tars, gasoline, grease, and a variety of other chemicals, making them suitable for demanding environments.
  • Therm-O-Rad sealed seams ensure waterproof protection
  • Premium snap lock suspender buckles for secure fit
  • Heavy-duty suspenders with crossover design
  • Heavy-duty snaps resist failure
  • Mildew resistant 2 mil exterior coating
  • Exterior coating provides snag and abrasion resistance
  • Sleek coating is easy to decontaminate and sheds liquids effectively
  • Heat-sealed seams are stronger than the material itself, allowing for extensive laundering