Learn More
MSC Tingley Jacket, DuraScrim, Double-Coated PVC/Polyester, Flame Resistant, Waterproof, 1 ea
Catalog No. 5000674684
Encompass
The DuraScrim Jacket is a double-coated PVC on polyester jacket offering high tear strength and durability. Designed for light chemical, construction, and utilities applications, it provides chemical resistance to various acids, oils, alcohols, salts, and alkalis.
  • Full cut shoulder with 90° sleeve-to-body angle for freedom of movement
  • Storm fly front seals out wind, rain, and contaminants
  • Hood snaps compatible with DuraScrim hood
  • Self-material collar resists contamination and has take-up snaps on sleeves
  • Non-conductive, non-corrosive hardware
  • Double-coated PVC on polyester for abrasion resistance
  • Mildew resistant
  • ASTM F1671 resistance to viral penetration
  • Therm-O-Rad sealed seams for 100% waterproof and chemical splash protection
  • Flame resistant tested per ASTM D6413
Catalog No. 50-006-74684 Supplier MSC Supplier No. 58661984
The DuraScrim Jacket is a double-coated PVC on polyester jacket offering high tear strength and durability. Designed for light chemical, construction, and utilities applications, it provides chemical resistance to various acids, oils, alcohols, salts, and alkalis.
  • Full cut shoulder with 90° sleeve-to-body angle for freedom of movement
  • Storm fly front seals out wind, rain, and contaminants
  • Hood snaps compatible with DuraScrim hood
  • Self-material collar resists contamination and has take-up snaps on sleeves
  • Non-conductive, non-corrosive hardware
  • Double-coated PVC on polyester for abrasion resistance
  • Mildew resistant
  • ASTM F1671 resistance to viral penetration
  • Therm-O-Rad sealed seams for 100% waterproof and chemical splash protection
  • Flame resistant tested per ASTM D6413