Learn More
MSC Tingley Jacket, Flame Resistant, PVC/Polyester, Hood Snaps, 1 ea.
Catalog No. 5000782243
Encompass
The Tingley DuraScrim Jacket provides superior protection for demanding work environments. Constructed from double-coated PVC on polyester, this flame-resistant jacket offers durability and reliable performance.
  • Flame resistant (ASTM D6413)
  • Waterproof with Therm-O-Rad sealed seams
  • Excellent chemical splash and abrasion resistance
  • Non-conductive, non-corrosive hardware
  • Mildew resistant
  • Full cut shoulder for maximum freedom of movement
  • Features hood snaps
Catalog No. 50-007-82243 Supplier MSC Supplier No. 58662057
The Tingley DuraScrim Jacket provides superior protection for demanding work environments. Constructed from double-coated PVC on polyester, this flame-resistant jacket offers durability and reliable performance.
  • Flame resistant (ASTM D6413)
  • Waterproof with Therm-O-Rad sealed seams
  • Excellent chemical splash and abrasion resistance
  • Non-conductive, non-corrosive hardware
  • Mildew resistant
  • Full cut shoulder for maximum freedom of movement
  • Features hood snaps